1,2,3,5-Tetra-O-acetyl-β-L-ribofuranose 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2R,3S,4S,5S)-5-(Acetoxymethyl)tetrahydrofuran-2,3,4-triyl triacetate
J66074-06
J66074-09
J66074-03
10814-408EA
43.92
USD
10814-408
10814-412
10814-410
1,2,3,5-Tetra-O-acetyl-β-L-ribofuranose 98%
1,2,3,5-Tetra-O-acetyl-beta-L-ribofuranose
Formula:
C₁₃H₁₈O₉ MW: 318.278 g/mol Melting Pt: 80…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00674547 CAS Number: 144490-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...