Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Protein Kinase C-delta Peptide Substrate

Supplier: Alfa Aesar
J66073-EXD J66073-MCR
10814-404 10814-406
Protein Kinase C-delta Peptide Substrate
Molecular Formula: C109H191N35O29S
Formula Weight: 2487.99
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty