N-Acetyl Cholecystokinin

Supplier: Thermo Scientific
J66056-MCR J66056-EXD
10815-434EA 70.68 USD
10815-434 10815-436
N-Acetyl Cholecystokinin
Proteins and Peptides
Molecular Formula: C33H41N7O12S2
Formula Weight: 791.85
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR