Caspase-8 Inhibitor I

Supplier: Thermo Scientific
J66050-EXD J66050-MCR
10815-422EA 227.21 USD
10815-422 10815-420
Caspase-8 Inhibitor I
Proteins and Peptides
Molecular Formula: C95H162N20O26
Formula Weight: 2000.41
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR