D-Glucuronamide 98%
Supplier: Thermo Scientific Chemicals
J66047-03
J66047-06
10815-416EA
83.5
USD
10815-416
10815-414
D-Glucuronamide 98%
D-Glucuronamide
Formula:
C₆H₁₁NO₆ Melting Pt: 166…167 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006619 CAS Number: 3789-97-7 EINECS: 223-260-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...