Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cecropin B, free acid

Supplier: Alfa Aesar
J66041-MA J66041-MCR J66041-LB0
10815-410EA 1119.16 USD InStock
10815-410 10815-412 10815-408
Cecropin B, free acid
Proteins and Peptides
Molecular Formula: C176H302N52O41S
Formula Weight: 3834.66
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty