[Tyr0] Urocortin

Supplier: Thermo Scientific
J66035-MCR
10815-406EA 385.3 USD
10815-406
[Tyr0] Urocortin
Proteins and Peptides
CAS No.: 187111-93-9
Molecular Formula: C215H347N63O66
Formula Weight: 4870.50
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00678221
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR