Growth Hormone Releasing Peptide-2, Thermo Scientific

Supplier: Thermo Scientific
J66029-MB J66029-LB0
10815-404EA 374.33 USD
10815-404 10815-402
Growth Hormone Releasing Peptide-2, Thermo Scientific
Proteins and Peptides
CAS No.: 158861-67-7
Molecular Formula: C42H50N8O5
Formula Weight: 746.90
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00674876
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR