Phenyl-4,6-O-benzylidene-β-D-glucopyranoside 98%
J66012-03
10815-370EA
44.92
USD
10815-370
Phenyl-4,6-O-benzylidene-β-D-glucopyranoside 98%
Phenyl-4,6-O-benzylidene-beta-D-glucopyranoside
Formula:
C₁₉H₂₀O₆ Melting Pt: 194…196 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00136366 CAS Number: 75829-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...