PPP

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (5R,5aS,8aR,9R)-9-Hydroxy-5-(3,4,5-trimethoxyphenyl)-5,8,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(5aH)-onePicropodophyllin

J65916-MA
10815-344EA 232.29 USD
10815-344
PPP
PPP

Picropodophyllotoxin is an insulin-like growth factor-I (IGF-I) receptor kinase inhibitor


  • Light Sensitive


White solid

Formula: C₂₂H₂₂O₈
MW: 414.40 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01742647
CAS Number: 477-47-4
UN: 3462
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR