Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Z-Ile-Glu(OMe)-Thr-Asp(OMe)-fluoromethyl ketone

Supplier: Alfa Aesar
J65505-MCR
10815-322EA 238.22 USD InStock
10815-322
Z-Ile-Glu(OMe)-Thr-Asp(OMe)-fluoromethyl ketone
Proteins and Peptides
Molecular Formula: C30H43FN4O11
Formula Weight: 654.68
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White
Solubility: Soluble in DMSO
MDL No.: MFCD01861686

An irreversible and cell permeable inhibitor of caspase-8 and granzyme B; also inhibits caspase-10.

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty