Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Transforming Growth Factor-beta 1, human platelets, carrier free, 97%, BT-213CF

Supplier: Alfa Aesar
J65044-YAM
10814-366EA 646.53 USD InStock
10814-366
Transforming Growth Factor-beta 1, human platelets, carrier free, 97%, BT-213CF
Proteins and Peptides
Purity: 97%
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
UN No.: 1993
MDL No.: MFCD00166089

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty