Would you like to log in again?
Cyclic AMP and GMP Enzyme Immunoassay Kit, Alfa Aesar
Supplier: Alfa Aesar
J64184-QA J65422-QA J64209-QA J64209-QA J64184-QA J65001-QA
CAAAJ64184-QA 10814-394 AAJ64209-QA CAAAJ64209-QA AAJ64184-QA 10814-362
Cyclic AMP and GMP Enzyme Immunoassay Kit, Alfa Aesar
Assays Enzyme Assays
Cyclic AMP and Cyclic GMP EIA kits are complete systems
  • Highly sensitive
  • Dual range
  • Non-radioactive
Levels of cAMP or cGMP can be conveniently measured from most biological samples (urine, plasma and cell culture) without the extensive purification procedures inherent in other methodologies. Both the Cyclic AMP and Cyclic GMP assays can be run either overnight or in 6 hours.
Each kit uses alkaline phosphatase labeled tracer (cAMP or cGMP) competing for sites on the antibody with standard or sample. Both assays employ a highly specific antibody to ensure accurate, consistent results.
Caution: Store in a cool place. Incompatible with strong oxidizing agents.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty