Would you like to log in again?
Osteocalcin, Human, Enzyme Immunoassay Kit, Alfa Aesar
Supplier: Alfa Aesar
J64764-QA J64816-QA
10814-338EA 404.48 USD InStock
10814-338 10814-340
Osteocalcin, Human, Enzyme Immunoassay Kit, Alfa Aesar
Assays Enzyme Assays
Osteocalcin is used as a bio-marker for the bone formation process and is involved to study the effectiveness of a glycoprotein on bone formation used osteocalcin as a measure of osteoblast activity.
  • Soluble in water
This osteocalcin is compatible with cultured cell extracts or scaffold cells. It acts as a hormone in the body, causing beta cells in the pancreas to release more insulin. Further, it is used for the quantitative determination of intact human osteocalcin in serum and plasma samples using 96-well plates and a microplate reader.
Caution: Incompatible with strong oxidizing agents.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty