Platelet-poor Plasma Derived Serum, bovine, BT-214

J64483-AB J64483-AE
10814-330EA 98.28 USD
10814-330 10814-332
Platelet-poor Plasma Derived Serum, bovine, BT-214
Platelet-poor Plasma Derived Serum, bovine is used in platelet aggregation studies to both adjust the Platelet-rich plasma concentration as well as to serve as a control

  • Physical Form: Liquid
  • MDL No.: MFCD01114289

It finds application in clinical studies such as blood management, orthopedic surgery and cosmetic surgery.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR