Would you like to log in again?
Mouse Osteocalcin Enzyme Immunoassay Kit, BT-470, Alfa Aesar
Supplier: Alfa Aesar
J64239-QA
10814-322
Mouse Osteocalcin Enzyme Immunoassay Kit, BT-470, Alfa Aesar
Mouse Osteocalcin Enzyme Immunoassay Kit is a sandwich enzyme immunoassay used in vitro quantitative measurement of mouse osteocalcin (OC in mouse) serum, plasma and other biological fluids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty