Benzyldimethyltetradecylammonium chloride hydrate 98%

Supplier: Alfa Aesar
Danger

J63465-14 J63465-14 J63465-06
AAJ63465-14EA 48.88 USD
CAAAJ63465-14 AAJ63465-14 10814-286
Benzyldimethyltetradecylammonium chloride hydrate 98%
Benzyldimethyltetradecylammonium chloride hydrate

Phase transfer catalyst

Formula: C₂₃H₄₂ClN·H₂O
MW: 368,05 (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00011771
CAS Number: 139-08-2
EINECS: 205-352-0
UN: 2923
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR