Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Transferrin (Apo), bovine plasma, 98+%

Supplier: Alfa Aesar
J61626-MC
10814-258EA 22.19 USD InStock
10814-258
Transferrin (Apo), bovine plasma, 98+%
Proteins and Peptides
Purity: 98+%
CAS No.: 11096-37-0
Storage Temperature: 2°C to 8°C
Physical Form: Lyophilized powder
Appearance: Off-white
Merck Reference: 14,9571
MDL No.: MFCD00130536

Blood plasma protein for iron delivery. Apo-transferrin is iron-free

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty