Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glutamic oxalacetic transaminase, porcine heart

Supplier: Alfa Aesar
J60044-JPL
10813-632EA 146.85 USD InStock
10813-632
Glutamic oxalacetic transaminase, porcine heart
Enzymes
CAS No.: 9000-97-9
Physical Form: Ammonium sulfate suspension
Appearance: Yellow
MDL No.: MFCD00131191

Converts L-aspartate and alpha-ketoglutarate to oxaloacetate and L-glutamate

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty