1,3,5-Triaza-7-phosphaadamantane ≥97%
Supplier: Thermo Scientific Chemicals
Synonyms:
PTA
H66785-06
H66785-03
AAH66785-06EA
393.99
USD
AAH66785-06
AAH66785-03
1,3,5-Triaza-7-phosphaadamantane ≥97%
1,3,5-Triaza-7-phosphaadamantane
Formula:
C₆H₁₂N₃P MW: 157.157 g/mol Melting Pt: 244…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00154905 CAS Number: 53597-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...