3-Iodoindole 96%

Warning

H66490-03 H66490-MD
AAH66490-03EA 274.15 USD
AAH66490-03 AAH66490-MD
3-Iodoindole 96%
3-Iodoindole
Formula: C₈H₆IN
Storage Temperature: Freezer
MDL Number: MFCD07778364
CAS Number: 26340-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR