3-Cyano-4-hydroxybenzeneboronic acid pinacol ester 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Hydroxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
H66434-MD
AAH66434-MDEA
134.99
USD
AAH66434-MD
3-Cyano-4-hydroxybenzeneboronic acid pinacol ester 95%
3-Cyano-4-hydroxybenzeneboronic acid pinacol ester
Formula:
C₁₃H₁₆BNO₃ MW: 245.085 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16994355 CAS Number: 775351-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...