Bis(acetonitrile)(1,5-cyclooctadiene)iridium(I) tetrafluoroborate ≥97%

Warning

H66412-03 H66412-MD
AAH66412-MDEA 83.29 USD
AAH66412-MD AAH66412-03
Bis(acetonitrile)(1,5-cyclooctadiene)iridium(I) tetrafluoroborate ≥97%
Bis(acetonitrile)(1,5-cyclooctadiene)iridium(I) tetrafluoroborate
Formula: C₁₂H₁₈BF₄IrN₂
Storage Temperature: Ambient
MDL Number: MFCD28144566
CAS Number: 32679-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR