5-(Boc-amino)pyridine-3-boronic acid pinacol ester 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-Butyl (5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)carbamate

H66223-MD
AAH66223-MDEA 278.99 USD
AAH66223-MD
5-(Boc-amino)pyridine-3-boronic acid pinacol ester 95%
5-(Boc-amino)pyridine-3-boronic acid pinacol ester
Formula: C₁₆H₂₅BN₂O₄
MW: 320.196 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13195277
CAS Number: 1171897-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR