6-Fluoro-2,3-dimethoxybenzoic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

H66209-MD
AAH66209-MDEA 246.99 USD
AAH66209-MD
6-Fluoro-2,3-dimethoxybenzoic acid 95%
6-Fluoro-2,3-dimethoxybenzoic acid
Formula: C₉H₉FO₄
Storage Temperature: Ambient
MDL Number: MFCD13193706
CAS Number: 265670-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR