Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥97%

Warning

Synonyms: Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I)tetrafluoroborate

H66191-03 H66191-06
AAH66191-03EA 693.99 USD
AAH66191-03 AAH66191-06
Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥97%
Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Formula: C₁₂H₁₈BF₄N₂Rh
MW: 293,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798085
CAS Number: 32679-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR