2-Chloro-N,N-dimethyl-4-nitroaniline 96%

Warning

H63488-06 H63488-03 H63488-MD
AAH63488-06EA 289.71 USD
AAH63488-06 AAH63488-MD AAH63488-03
2-Chloro-N,N-dimethyl-4-nitroaniline 96%
2-Chloro-N,N-dimethyl-4-nitroaniline
Formula: C₈H₉ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00043604
CAS Number: 6213-19-0
EINECS: 228-280-1
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR