3-Bromo-5-fluorophenylacetonitrile 96%

Warning

Synonyms: 2-(3-bromo-5-fluorophenyl)acetonitrileBenzeneacetonitrile, 3-bromo-5-fluoro-2-(3-bromo-5-fluorophenyl)ethanenitrile

H62605-MD H62605-06 H62605-03
AAH62605-03EA 174.27 USD
AAH62605-03 AAH62605-06 AAH62605-MD
3-Bromo-5-fluorophenylacetonitrile 96%
3-Bromo-5-fluorophenylacetonitrile
Formula: C₈H₅BrFN
MW: 214.037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258960
CAS Number: 305800-58-2
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR