3-Bromo-2-cyanobenzeneboronic acid 96%

Warning

Synonyms: (3-Bromo-2-cyanophenyl)boronic acid

H61871-03 H61871-MD
AAH61871-MDEA 44.57 USD
AAH61871-MD AAH61871-03
3-Bromo-2-cyanobenzeneboronic acid 96%
3-Bromo-2-cyanobenzeneboronic acid
Formula: C₇H₅BBrNO₂
MW: 225.836 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10696672
CAS Number: 1032231-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR