Potassium-2-methoxypyridine-5-trifluoroborate 97%

Warning

H61645-06 H61645-03
AAH61645-06EA 233.81 USD
AAH61645-06 AAH61645-03
Potassium-2-methoxypyridine-5-trifluoroborate 97%
Potassium-2-methoxypyridine-5-trifluoroborate
Formula: C₆H₆BF₃KNO
Storage Temperature: Ambient
MDL Number: MFCD09993111
CAS Number: 1144573-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR