6-Bromo-3,4-dihydro-2H-1,4-benzoxazine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Bromo-3,4-dihydro-2H-benzo[b][1,4]oxazine

H61070-MD H61070-03
AAH61070-MDEA 78.79 USD
AAH61070-MD AAH61070-03
6-Bromo-3,4-dihydro-2H-1,4-benzoxazine 97%
6-Bromo-3,4-dihydro-2H-1,4-benzoxazine
Formula: C₈H₈BrNO
MW: 214.062 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08544341
CAS Number: 105655-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR