7-Bromo-3,4-dihydro-2H-1,4-benzothiazine 97%

Supplier: Thermo Scientific Chemicals
Warning

H61021-03 H61021-MD
AAH61021-03EA 267.99 USD
AAH61021-03 AAH61021-MD
7-Bromo-3,4-dihydro-2H-1,4-benzothiazine 97%
7-Bromo-3,4-dihydro-2H-1,4-benzothiazine
Formula: C₈H₈BrNS
Storage Temperature: Ambient
MDL Number: MFCD11847754
CAS Number: 193414-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR