1-Fluoro-2-ethynylbenzene 97%

Supplier: Alfa Aesar
Warning

Synonyms: (2-Fluorophenyl)acetylene (2-Fluorophenyl)ethyne 1-Ethynyl-2-fluorobenzene

H60596-03 H60596-MD
AAH60596-03EA 124.44 USD
AAH60596-03 AAH60596-MD
1-Fluoro-2-ethynylbenzene 97%
1-Fluoro-2-ethynylbenzene
Formula: C₈H₅F
MW: 120.13 g/mol
Boiling Pt: 150 °C
Density: 1,060
Flash Pt: 37 °C (99 °F)
Storage Temperature: Ambient
MDL Number: MFCD00799540
CAS Number: 766-49-4
EINECS: 000-000-0
UN: 1993
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR