(R)-2-Amino-1-diphenylphosphino-3,3-dimethylbutane ≥97%
|
Danger
|
Synonyms:
(R)-1-(Diphenylphosphino)-3,3-dimethyl-2-butylamine
H60508-MD
H60508-03
AAH60508-03EA
319.68
USD
AAH60508-03
AAH60508-MD
(R)-2-Amino-1-diphenylphosphino-3,3-dimethylbutane ≥97%
(R)-2-Amino-1-diphenylphosphino-3,3-dimethylbutane
Formula:
C₁₈H₂₄NP MW: 285.37 g/mol Density: 1.026 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD17013994 CAS Number: 1366384-12-4 UN: 2735 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...