2-Mercapto-4-(trifluoromethyl)pyrimidine 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Sulphanyl-4-(trifluoromethyl)pyrimidine

H60501-03 H60501-06
AAH60501-03EA 60.59 USD
AAH60501-03 AAH60501-06
2-Mercapto-4-(trifluoromethyl)pyrimidine 96%
2-Mercapto-4-(trifluoromethyl)pyrimidine
Formula: C₅H₃F₃N₂S
MW: 180.148 g/mol
Melting Pt: 164…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00115113
CAS Number: 136547-17-6
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR