1-Benzylbenzimidazole 97%

Warning

H60088-03 H60088-06
AAH60088-03EA 31.11 USD
AAH60088-03 AAH60088-06
1-Benzylbenzimidazole 97%
1-Benzylbenzimidazole
Formula: C₁₄H₂₁NO₂S
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00458798
CAS Number: 141437-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR