α,4-Dibromo-3-fluorotoluene 96%
|
Danger
|
Synonyms:
4-Bromo-3-fluorobenzyl bromide
, 1-Bromo-4-(bromomethyl)-2-fluorobenzene
H59275-03
H59275-MD
AAH59275-03EA
184.37
USD
AAH59275-03
AAH59275-MD
α,4-Dibromo-3-fluorotoluene 96%
α,4-Dibromo-3-fluorotoluene
Formula:
C₇H₅Br₂F MW: 267.92 g/mol Melting Pt: 33…36 °C Storage Temperature: Ambient |
MDL Number:
MFCD11847179 CAS Number: 127425-73-4 UN: 3261 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...