cyclohexylzinc bromide 0.5 M in THF

Supplier: Thermo Scientific Chemicals
Danger


Packaged under Argon in resealable ChemSeal™ bottles.

H58852-AD
AAH58852-ADEA 240.24 USD
AAH58852-AD
cyclohexylzinc bromide 0.5 M in THF
cyclohexylzinc bromide
Formula: C₆H₁₁BrZn
Density: 0.963
Storage Temperature: Ambient
MDL Number: MFCD00671993
CAS Number: 7565-57-3
UN: 3399
ADR: "4.3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR