Cyclopentylzinc bromide solution 0.5 M in THF

Supplier: Thermo Scientific Chemicals
Danger


Packaged under Argon in resealable ChemSeal™ bottles.

H58764-AD
AAH58764-ADEA 253 USD
AAH58764-AD
Cyclopentylzinc bromide solution 0.5 M in THF
Cyclopentylzinc bromide solution
Formula: C₅H₉BrZn
Density: 0.955
Storage Temperature: Ambient
MDL Number: MFCD00671991
CAS Number: 171860-68-7
UN: 3399
ADR: "4.3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR