4-Bromo-2-fluorobenzylzinc bromide 0.5 M in THF
|
Danger
|
Synonyms:
Bromo(4-bromo-2-fluorobenzyl)zinc
Packaged under Argon in resealable ChemSeal™ bottles.
H58746-AD
AAH58746-ADEA
239.84
USD
AAH58746-AD
4-Bromo-2-fluorobenzylzinc bromide 0.5 M in THF
4-Bromo-2-fluorobenzylzinc bromide
Formula:
C₇H₅Br₂FZn MW: 333.3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13152553 UN: 3399 ADR: "4.3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...