4-chlorophenylzinc iodide 0.5 M in THF

Danger


Packaged under Argon in resealable ChemSeal™ bottles.

H58740-AD
AAH58740-ADEA 192.55 USD
AAH58740-AD
4-chlorophenylzinc iodide 0.5 M in THF
4-chlorophenylzinc iodide
Formula: C₆H₄ClIZn
Density: 1.066
Storage Temperature: Ambient
MDL Number: MFCD01311392
CAS Number: 151073-70-0
UN: 3399
ADR: "4.3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR