4-methoxyphenylzinc iodide 0.5 M in THF

Danger


Packaged under Argon in resealable ChemSeal™ bottles.

H58690-AD
AAH58690-ADEA 205.09 USD
AAH58690-AD
4-methoxyphenylzinc iodide 0.5 M in THF
4-methoxyphenylzinc iodide
Formula: C₇H₇IOZn
Density: 1.006
Storage Temperature: Ambient
MDL Number: MFCD01311444
CAS Number: 254454-47-2
UN: 3399
ADR: "4.3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR