6-Cyano-2-(2-pyridyl)benzimidazole 97%

Warning

Synonyms: 2-(pyridin-2-yl)-1H-1,3-benzodiazole-5-carbonitrile2-(pyridin-2-yl)-1H-benzo[d]imidazole-5-carbonitrile

H58615-03 H58615-06
AAH58615-03EA 245.14 USD
AAH58615-03 AAH58615-06
6-Cyano-2-(2-pyridyl)benzimidazole 97%
6-Cyano-2-(2-pyridyl)benzimidazole
Formula: C₁₃H₈N₄
MW: 220.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22581244
CAS Number: 226702-84-7
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR