n-Nonylzinc bromide 0.5 M in THF

Supplier: Thermo Scientific Chemicals
Danger


Packaged under Argon in resealable ChemSeal™ bottles.

H58604-AD
AAH58604-ADEA 179.99 USD
AAH58604-AD
n-Nonylzinc bromide 0.5 M in THF
n-Nonylzinc bromide
Formula: C₉H₁₉BrZn
Storage Temperature: Ambient
MDL Number: MFCD09801447
CAS Number: 626207-32-7
UN: 3399
ADR: "4.3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR