3-Phenylpropylzinc bromide 0.5 M in THF

Supplier: Thermo Scientific Chemicals
Danger


Packaged under Argon in resealable ChemSeal™ bottles.

H58569-AD
AAH58569-ADEA 224.99 USD
AAH58569-AD
3-Phenylpropylzinc bromide 0.5 M in THF
3-Phenylpropylzinc bromide
Formula: C₉H₁₁BrZn
Storage Temperature: Ambient
MDL Number: MFCD08064069
CAS Number: 626207-42-9
UN: 3399
ADR: "4.3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR