3-Maleimidobenzonitrile 97%

Warning

Synonyms: 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzonitrile3-(2,5-dioxopyrrol-1-yl)benzonitrile

H58479-03 H58479-06
AAH58479-06EA 1047.78 USD
AAH58479-06 AAH58479-03
3-Maleimidobenzonitrile 97%
3-Maleimidobenzonitrile
Formula: C₁₁H₆N₂O₂
MW: 198.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00176056
CAS Number: 303770-14-1
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR