pyrazolo[1,5-a]pyrimidine-3-carbonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Cyanopyrazolo[1,5-a]pyrimidine

H58380-09 H58380-04
AAH58380-09EA 762.57 USD
AAH58380-09 AAH58380-04
pyrazolo[1,5-a]pyrimidine-3-carbonitrile 97%
pyrazolo[1,5-a]pyrimidine-3-carbonitrile
Formula: C₇H₄N₄
Storage Temperature: Ambient
MDL Number: MFCD01927799
CAS Number: 25939-87-1
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR