6-Methyl-2-pyridylzinc bromide 0.5 M in THF

Danger


Packaged under Argon in resealable ChemSeal™ bottles.

H58354-AD
AAH58354-ADEA 235.05 USD
AAH58354-AD
6-Methyl-2-pyridylzinc bromide 0.5 M in THF
6-Methyl-2-pyridylzinc bromide
Formula: C₆H₆BrNZn
Density: 0.996
Storage Temperature: Ambient
MDL Number: MFCD01311462
CAS Number: 308795-98-4
UN: 3399
ADR: "4.3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR