tert-Butylzinc bromide 0.5 M in THF

Supplier: Thermo Scientific Chemicals
Danger


Packaged under Argon in resealable ChemSeal™ bottles.

H58351-AD
AAH58351-ADEA 294.99 USD
AAH58351-AD
tert-Butylzinc bromide 0.5 M in THF
tert-Butylzinc bromide
Formula: C₄H₉BrZn
Density: 0.966
Storage Temperature: Ambient
MDL Number: MFCD00671989
CAS Number: 7565-59-5
UN: 3399
ADR: "4.3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR