2-Methoxyphenylzinc iodide 0.5 M in THF

Danger


Packaged under Argon in resealable ChemSeal™ bottles.

H58341-AD
AAH58341-ADEA 167.37 USD
AAH58341-AD
2-Methoxyphenylzinc iodide 0.5 M in THF
2-Methoxyphenylzinc iodide
Formula: C₇H₇IOZn
Density: 0.997
Storage Temperature: Ambient
MDL Number: MFCD01311454
CAS Number: 282727-20-2
UN: 3399
ADR: "4.3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR